Learn More
Value Plastics Female Luer Thread Style to NPT Adapter Fittings, Straight
Luer fittings allow for a simple yet secure connection
608.24 NOK - 795.99 NOK
Product Code | Brand | Material (Seal) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material (Seal) | Price | Quantity & Availability | ||||||
30053952
|
Value Plastics
50119-95 |
Prolypropelene |
795.99 NOK
Pack of 25 |
Estimated Shipment: 21-05-2025
Log in to see stock. |
||||||
30052983
|
Value Plastics
50119-94 |
White Nylon |
608.24 NOK
Pack of 25 |
Estimated Shipment: 21-05-2025
Log in to see stock. |
||||||
Description
Quick and easy assembly and disassembly without the need for tools. Thread design provides a secure closure—preventing contamination. Conforms to ISO-594-1 and ISO 594-2 standardsSpecifications
1/8-27 NPT(M) Thread |
Product Title
Value Plastics Female Luer Thread Style to NPT Adapter Fittings, Straight