Learn More
Value Plastics Female Luer Thread Style to NPT Adapter Fittings, Straight
Luer fittings allow for a simple yet secure connection
Brand:  Value Plastics 50119-95
 View more versions of this product
Product Code. 30053952
  • 795.99 NOK / Pack of 25
Estimated Shipment: 10-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Quick and easy assembly and disassembly without the need for tools. Thread design provides a secure closure—preventing contamination. Conforms to ISO-594-1 and ISO 594-2 standards
Specifications
Specifications
ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
1/8-27 NPT(M) Thread
Prolypropelene
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Value Plastics Female Luer Thread Style to NPT Adapter Fittings, Straight > Prolypropelene