Learn More
Falcon™ Polystyrene Aspirating Pipets, Without Graduations
Manufactured from polystyrene as a safer alternative to glass Pasteur pipets. Falcon™ Disposable Aspirating Pipets are ideal for tissue culture usage or any non-toxic solution that requires aliquoted or aspirating/dispensing in a sterile environment.
Brand: Falcon™ 357501
614.49 NOK valid until 2025-08-24
Use promo code "24140" to get your promotional price.
Description
- Sterile, polystyrene, non-plugged, non-graduated pipets
- Safer alternative to glass Pasteur pipets
- Can be used for all vacuum-aspirating procedures
- Noncytotoxic, nonpyrogenic tested to less than 0.1 EU/mL
- Individually packaged in thermoformed paper-plastic wrap to ensure sterile presentation
Order Info
All sizes come individually wrapped, 200 per case
Specifications
Class I Medical | |
Polystyrene | |
For tissue culture usage or any non-toxic solution that requires aliquot or aspirating, dispensing in a sterile environment | |
5 mL | |
Single-use |
Aspirating | |
Sterile | |
13.71 in. | |
Aspirating Pipet |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
4308716 | Certificate of Quality | 21/05/2025 | 357501 |
4214018 | Certificate of Quality | 25/12/2024 | 357501 |
3344299 | Certificate of Compliance | 08/02/2024 | 357501 |
3002008 | Certificate of Compliance | 21/07/2023 | 357501 |
2335015 | Certificate of Compliance | 27/05/2023 | 357501 |
Promotions
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
New Offer Save Up to 59%