Learn More
Corning™ Costar™ Disposable Aspirating Pipets
GREENER_CHOICE
Constructed of polystyrene for strength, clarity, and durability. Corning™ Costar™ Disposable Aspirating Pipets are ideal for aseptically aspirating liquids by using vacuum suction.
Brand:  Corning™ 9099
1607.85 NOK valid until 2025-08-24
Use promo code "24140" to get your promotional price.
 View more versions of this product
Product Code. 10424342
  • 1764.98 NOK / Pack of 200
Quality Analysis and Quality Control Testing
Use promo code 24140 at checkout
View Available Promotions
Only null left
Estimated Shipment: 12-09-2025
Description
  • One piece polystyrene construction
  • Sterile and certified nonpyrogenic
  • No graduations; unplugged
  • Tip shape: round
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Aspirating
13.7 in.
Individual Clear Plastic Wrap
5 mL
Single-use
Polystyrene
Round
0.169 oz.
34.8 cm
Documents
Product Information
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number08921029 Certificate TypeCertificate of Compliance Date15/07/2021 Catalog Number9099
Lot Number06321030 Certificate TypeCertificate of Compliance Date15/07/2021 Catalog Number9099
Lot Number12220053 Certificate TypeCertificate of Compliance Date01/07/2021 Catalog Number9099
Lot Number06021043 Certificate TypeCertificate of Compliance Date01/07/2021 Catalog Number9099
Promotions
Promotions
Product Content Correction
Product Title
Corning™ Costar™ Disposable Aspirating Pipets > Capacity: 5mL; 200/cs