missing translation for 'onlineSavingsMsg'
Learn More

EXT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1696.32 NOK - 4796.50 NOK

Specifications

Antigen EXT2
Concentration 0.14 mg/mL
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16848363
View Documents
Proteintech
11348-1-AP-20UL
20 μL
1696.32 NOK
20µL
Please to purchase this item. Need a web account? Register with us today!
16838363
View Documents
Proteintech
11348-1-AP-150UL
150 μL
4796.50 NOK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This gene encodes one of two glycosyltransferases involved in the chain elongation step of heparan sulfate biosynthesis. Mutations in this gene cause the type II form of multiple exostoses. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.
Specifications

Specifications

EXT2
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human, Mouse, Rat
P70428, Q93063
14043, 2132, 311215
EXT2 Fusion Protein Ag1748
Primary
-20°C
EXT2
0.14 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
exostoses (multiple) 2, Exostosin 2, EXT2, Multiple exostoses protein 2, SOTV
EXT2
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.