missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EXT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11348-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes one of two glycosyltransferases involved in the chain elongation step of heparan sulfate biosynthesis. Mutations in this gene cause the type II form of multiple exostoses. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.Specifications
EXT2 | |
Polyclonal | |
Unconjugated | |
EXT2 | |
exostoses (multiple) 2, Exostosin 2, EXT2, Multiple exostoses protein 2, SOTV | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14043, 2132, 311215 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P70428, Q93063 | |
EXT2 | |
EXT2 Fusion Protein Ag1748 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction