Learn More
Thermo Scientific™ Eutech™ Replacement pH/Conductivity/Temp Sensor
Optimize your Eutech™ pH meters with these accessories and replacement parts.
Brand:  Thermo Scientific™ 01X097108
720.23 NOK valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Product Code. 10553484
  • 1089.90 NOK / Each
In Stock
to see stock.
Explore more special offers
Description
Optimize your Eutech™ meters with these accessories and replacement parts. Eutech™ Replacement Sensor is a replacement pH/Conductivity/Temp Sensor for PCTestr 35, PTTestr 35, PCSTestr 35.
Replacement pH/Conductivity/Temp sensor for:
  • PCTestr 35
  • PTTestr 35
  • PCSTestr 35
  • TRUSTED_SUSTAINABILITY
Specifications
Specifications
Replacement Part
Product Title
Thermo Scientific™ Eutech™ Replacement pH/Conductivity/Temp Sensor >