Learn More
3M™ Powered Air Respirator System Starter Kit
Powered Air Respirator System Starter Kit is a durable powered air respirator system designed for heavy duty environments at an affordable price. This starter kit includes a turbo unit, battery, charger and belt.
Brand: 3M™ 7100299964
12243.29 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- For a durable, economical powered air respirator starter kit use 3M™ Powered Air Respirator System Starter Kit PF-619E.
- This kit includes a 3M™ PF-600E turbo unit, charger and belt.
- The 3M™ PF-600E turbo unit features audible and visual alarms that alert the user to low battery or if the flow rate falls below the manufacturer's minimum design flow rate of 160 l/min.
- The unit incorporates a simple button operation, and the button is shielded to prevent accidental shut-off.
- It's designed to be compatible with loose fitting 3M™Versaflo™ M-200 and M-300 range of helmets, tight fitting 3M™ Promask™ FM3 and 3M™ Vision FM4 Full facepieces and the 3M™ DT-Series range of filters.
- The unit has an IP 65 rating with filters and decontamination plugs fitted to the filter and hose ports, making it suitable for cleaning by showering/water spray.
- The integrated battery is safely enclosed within the turbo unit casing and is rechargeable in-situ.
Specifications
White | |
Various | |
Standard |
Reusable | |
PAPR Kit |
Product Content Correction
Product Title
3M™ Powered Air Respirator System Starter Kit > Air Respirator