Learn More
Lenz Laborglasinstrumente™ Erlenmeyer flask with standard ground joint
Made of borosilicate glass. Lenz Laborglasinstrumente™ Erlenmeyer flask with standard ground joint are excellent for use in the laboratory for measuring chemicals or boiling liquids.
2485.00 NOK - 2735.00 NOK
Specifications
Material Borosilicate Glass
Graduated Yes
View More Specs
Products 3
Product Code Brand Capacity (Metric) Socket Joint Price Quantity & Availability  
Product Code Brand Capacity (Metric) Socket Joint Price Quantity & Availability  
10652852
View Documents
Lenz Laborglasinstrumente™
03024546
200 mL 45/40
2555.00 NOK
Pack of 10
Estimated Shipment: 02-05-2025
to see stock.
Add to basket
15350885
View Documents
Lenz Laborglasinstrumente™
03022470
1000 mL 24/29
2485.00 NOK
Pack of 10
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
10580122
View Documents
Lenz Laborglasinstrumente™
03024576
3000 mL 45/40
2735.00 NOK
Pack of 2
Estimated Shipment: 29-04-2025
Description
  • According to DIN EN ISO 4797
  • With graduation for reading the approximate volume
  • With labeling field
Specifications
Specifications
Borosilicate Glass
Yes
Documents
Product Certifications
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Erlenmeyer flask with standard ground joint