Learn More
Lenz Laborglasinstrumente™ Erlenmeyer flask with standard ground joint
Made of borosilicate glass. Lenz Laborglasinstrumente™ Erlenmeyer flask with standard ground joint are excellent for use in the laboratory for measuring chemicals or boiling liquids.
Brand: Lenz Laborglasinstrumente™ 03024576
This item is not returnable.
View return policy
Description
- According to DIN EN ISO 4797
- With graduation for reading the approximate volume
- With labeling field
Specifications
DIN EN ISO 4797 | |
Borosilicate Glass | |
45/40 | |
185 mm |
3000 mL | |
45/40 | |
Yes |