Learn More
Lenz Laborglasinstrumente™ Erlenmeyer flask with standard ground joint
Made of borosilicate glass. Lenz Laborglasinstrumente™ Erlenmeyer flask with standard ground joint are excellent for use in the laboratory for measuring chemicals or boiling liquids.
Brand:  Lenz Laborglasinstrumente™ 03024576
 View more versions of this product
Product Code. 10580122
  • 2735.00 NOK / Pack of 2
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • According to DIN EN ISO 4797
  • With graduation for reading the approximate volume
  • With labeling field
Specifications
Specifications
DIN EN ISO 4797
Borosilicate Glass
45/40
185 mm
3000 mL
45/40
Yes