missing translation for 'onlineSavingsMsg'
Learn More

EMX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1696.32 NOK - 4796.50 NOK

Specifications

Antigen EMX1
Concentration 0.36 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16856475
View Documents
Proteintech
55032-1-AP-20UL
20 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16846475
View Documents
Proteintech
55032-1-AP-150UL
150 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

EMX1 is a transcription factor, which in cooperation with EMX2. It acts to generate the boundary between the roof and archipallium in the developing brain. It may function in combinations with OTX1/2 to specify cell fates in the developing central nervous system.
Specifications

Specifications

EMX1
Western Blot
Unconjugated
Rabbit
Mouse, Human, Rat
Q04741, Q04742
13796, 2016, 500235
Peptide
Primary
-20°C
EMX1
0.36 mg/mL
Polyclonal
Liquid
RUO
0.1 M glycine, 0.1 M sodium bicarbonate with 50% glycerol and 0.02% sodium azide; pH 7.3
empty spiracles homeobox 1, Empty spiracles homolog 1, Empty spiracles like protein 1, EMX1, Homeobox protein EMX1
Emx1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.