missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EMX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55032-1-AP-150UL
This item is not returnable.
View return policy
Description
EMX1 is a transcription factor, which in cooperation with EMX2. It acts to generate the boundary between the roof and archipallium in the developing brain. It may function in combinations with OTX1/2 to specify cell fates in the developing central nervous system.Specifications
EMX1 | |
Polyclonal | |
Unconjugated | |
EMX1 | |
empty spiracles homeobox 1, Empty spiracles homolog 1, Empty spiracles like protein 1, EMX1, Homeobox protein EMX1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13796, 2016, 500235 | |
-20°C | |
Liquid |
Western Blot | |
0.36 mg/mL | |
0.1 M glycine, 0.1 M sodium bicarbonate with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q04741, Q04742 | |
Emx1 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction