missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGFR Monoclonal Antibody (199.12), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA513319
This item is not returnable.
View return policy
Description
MA5-13319 targets Epidermal Growth Factor Receptor in IP, ICC/IF and WB applications and shows reactivity with Human samples. The MA5-13319 immunogen is extracellular domain of human recombinant EGFR protein.
EGFR Monoclonal antibody specifically detects EGFR in Human samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blot.
Specifications
EGFR | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P00533 | |
EGFR | |
Extracellular domain of human recombinant EGFR protein | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
199.12 | |
Unconjugated | |
EGFR | |
2.7.10.1; 9030024J15Rik; AI552599; avian erythroblastic leukemia viral (v-erbB) oncogene homolog; avian erythroblastic leukemia viral (v-erb-b) oncogene homolog; cell growth inhibiting protein 40; cell proliferation-inducing protein 61; EC 2.7.10.1; egf receptor; Egfr; EGFR-related peptide; epidermal growth factor receptor; epidermal growth factor receptor (erythroblastic leukemia viral (v-erb-b) oncogene homolog, avian); Epidermal growth factor receptor formerly avian erythroblastic leukemia viral (v-erbB) oncogene homolog (Erbb1); epidermal growth factor receptor, formerly avian erythroblastic leukemia viral (v-erbB) oncogene homolog (Erbb1); ERBB; ERBB1; ErbB-1; erb-b2 receptor tyrosine kinase 1; Errb1; Errp; HER1; kinase EGFR; mENA; NISBD2; Oncogene ERBB; PIG61; Proto-oncogene c-ErbB-1; receptor tyrosine-protein kinase erbB-1; Urogastrone; wa2; wa-2; Wa5; waved 2 | |
Mouse | |
Protein A | |
RUO | |
1956 | |
4° C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
EGFR Monoclonal Antibody (199.12), Invitrogen™ > 500 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction