missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Eco AluRack for Upright Freezers

Providing a whole new way of making storage racks. Tenak™ Eco AluRacks for Upright Freezers fit into any freezer and can be used with all standard cryoboxes.

Brand:  Tenak™ TE11286

 View more versions of this product

Product Code. 12472540

  • 1270.00 NOK / Each
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Made of seawater resistant aluminum
  • Available in a range of box heights
  • Racks can be used with all freezer brands
  • Sold without cryoboxes
Specifications

Specifications

Eco AluRack
Boxes (15)
15
Yes
1 Pack
683 x 139mm
140 mm
696 mm
3 x 5
Aluminum
15
For Mtp, chest freezers
1 Pack
688 x 139 x 240mm
240 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers