missing translation for 'onlineSavingsMsg'
Learn More

E-64, ∽99%, MP Biomedicals™

853.30 NOK - 4997.90 NOK

Chemical Identifiers

CAS 66701-25-5
Molecular Formula C15H27N5O5
Molecular Weight (g/mol) 357.41
MDL Number MFCD00080261,MFCD00080261
InChI Key LTLYEAJONXGNFG-HBNTYKKESA-N
Synonym d09fgq, l-3-trans-carboxy-oxiran-2-carbonyl-leu-agmatin, trans-epoxysuccinyl-leucylamido-4-guanidino butane, 3-1-4-carbamimidamidobutyl carbamoyl-3-methylbutyl carbamoyl oxirane-2-carboxylic acid, 3-1-4-diaminomethylideneamino butylamino-4-methyl-1-oxopentan-2-yl carbamoyl oxirane-2-carboxylic acid
PubChem CID 4641099
IUPAC Name (2R,3R)-3-{[(1S)-1-({4-[(diaminomethylidene)amino]butyl}carbamoyl)-3-methylbutyl]carbamoyl}oxirane-2-carboxylic acid
SMILES CC(C)C[C@H](NC(=O)[C@@H]1O[C@H]1C(O)=O)C(=O)NCCCCN=C(N)N
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11474001
View Documents Promotion Details
MP Biomedicals
0215284601
1 mg
853.30 NOK
1mg
Please to purchase this item. Need a web account? Register with us today!
11484001
View Documents Promotion Details
MP Biomedicals
0215284605
5 mg
2814.30 NOK
5mg
Please to purchase this item. Need a web account? Register with us today!
11494001
View Documents Promotion Details
MP Biomedicals
0215284610
10 mg
4997.90 NOK
10mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

66701-25-5
357.41
LTLYEAJONXGNFG-HBNTYKKESA-N
4641099
CC(C)C[C@H](NC(=O)[C@@H]1O[C@H]1C(O)=O)C(=O)NCCCCN=C(N)N
C15H27N5O5
MFCD00080261,MFCD00080261
d09fgq, l-3-trans-carboxy-oxiran-2-carbonyl-leu-agmatin, trans-epoxysuccinyl-leucylamido-4-guanidino butane, 3-1-4-carbamimidamidobutyl carbamoyl-3-methylbutyl carbamoyl oxirane-2-carboxylic acid, 3-1-4-diaminomethylideneamino butylamino-4-methyl-1-oxopentan-2-yl carbamoyl oxirane-2-carboxylic acid
(2R,3R)-3-{[(1S)-1-({4-[(diaminomethylidene)amino]butyl}carbamoyl)-3-methylbutyl]carbamoyl}oxirane-2-carboxylic acid

Specifications

66701-25-5
∽99%
MFCD00080261,MFCD00080261
LTLYEAJONXGNFG-HBNTYKKESA-N
(2R,3R)-3-{[(1S)-1-({4-[(diaminomethylidene)amino]butyl}carbamoyl)-3-methylbutyl]carbamoyl}oxirane-2-carboxylic acid
4641099
Powder
White
C15H27N5O5
d09fgq, l-3-trans-carboxy-oxiran-2-carbonyl-leu-agmatin, trans-epoxysuccinyl-leucylamido-4-guanidino butane, 3-1-4-carbamimidamidobutyl carbamoyl-3-methylbutyl carbamoyl oxirane-2-carboxylic acid, 3-1-4-diaminomethylideneamino butylamino-4-methyl-1-oxopentan-2-yl carbamoyl oxirane-2-carboxylic acid
CC(C)C[C@H](NC(=O)[C@@H]1O[C@H]1C(O)=O)C(=O)NCCCCN=C(N)N
357.41
357.4
E-64
Videos
SDS
Documents

Documents

Product Certifications