missing translation for 'onlineSavingsMsg'
Learn More

E-64, ∽99%, MP Biomedicals™

Product Code. 11484001
Click to view available options
Quantity:
1 mg
5 mg
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Identifiants chimiques

CAS 66701-25-5
Molecular Formula C15H27N5O5
Molecular Weight (g/mol) 357.41
MDL Number MFCD00080261,MFCD00080261
InChI Key LTLYEAJONXGNFG-HBNTYKKESA-N
Synonym d09fgq, l-3-trans-carboxy-oxiran-2-carbonyl-leu-agmatin, trans-epoxysuccinyl-leucylamido-4-guanidino butane, 3-1-4-carbamimidamidobutyl carbamoyl-3-methylbutyl carbamoyl oxirane-2-carboxylic acid, 3-1-4-diaminomethylideneamino butylamino-4-methyl-1-oxopentan-2-yl carbamoyl oxirane-2-carboxylic acid
PubChem CID 4641099
IUPAC Name (2R,3R)-3-{[(1S)-1-({4-[(diaminomethylidene)amino]butyl}carbamoyl)-3-methylbutyl]carbamoyl}oxirane-2-carboxylic acid
SMILES CC(C)C[C@H](NC(=O)[C@@H]1O[C@H]1C(O)=O)C(=O)NCCCCN=C(N)N

Spécification

Color White
Assay Percent Range ∽99%
Quantity 5 mg
Formula Weight 357.4
Physical Form Powder
Chemical Name or Material E-64
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'