Learn More
Lenz Laborglasinstrumente™ Extractor Heads Acc. to Soxhlet
For the extraction of liquids with specific light solvents. Lenz Laborglasinstrumente™ Extractor Heads Acc. to Soxhlet are manufactured from durable DURAN™ tubing.
827.00 NOK - 1835.00 NOK
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
10740612
|
Lenz Laborglasinstrumente™
05395137 |
100 mL |
1130.00 NOK
Each |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||||
10251741
|
Lenz Laborglasinstrumente™
05395149 |
250 mL |
1270.00 NOK
Each |
Estimated Shipment: 28-05-2025
Log in to see stock. |
||||||
10700612
|
Lenz Laborglasinstrumente™
05395125 |
30 mL |
827.00 NOK
Each |
Estimated Shipment: 10-06-2025
Log in to see stock. |
||||||
10392951
|
Lenz Laborglasinstrumente™
05395143 |
150 mL |
1190.00 NOK
Each |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
10662361
|
Lenz Laborglasinstrumente™
05395131 |
70 mL |
958.00 NOK
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
10343051
|
Lenz Laborglasinstrumente™
05395158 |
500 mL |
1835.00 NOK
Each |
Estimated Shipment: 03-07-2025
Log in to see stock. |
||||||
Description
- Acc to DIN 12602 and Soxhlet
- With cone NS 29/32
- Available with/without NS stopcock
- Corrosion-free and resistant to thermal shock
Specifications
Extractor Head | |
Duran Glass |
Product Title
Lenz Laborglasinstrumente™ Extractor Heads Acc. to Soxhlet