Learn More
Lenz Laborglasinstrumente™ Extractor Heads Acc. to Soxhlet
For the extraction of liquids with specific light solvents. Lenz Laborglasinstrumente™ Extractor Heads Acc. to Soxhlet are manufactured from durable DURAN™ tubing.
Brand:  Lenz Laborglasinstrumente™ 05395125
 View more versions of this product
Product Code. 10700612
  • 827.00 NOK / Each
Only null left
Estimated Shipment: 24-07-2025
Description
  • Acc to DIN 12602 and Soxhlet
  • With cone NS 29/32
  • Available with/without NS stopcock
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
Extractor Head
30 mL
Duran Glass
DIN 12602
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Extractor Heads Acc. to Soxhlet > Capacity: 30mL