Learn More
DL-Cysteine hydrochloride, 90+%, Thermo Scientific Chemicals
1377.00 NOK - 11853.00 NOK
Chemical Identifiers
CAS | 10318-18-0 |
---|---|
Molecular Formula | C3H8ClNO2S |
Molecular Weight (g/mol) | 157.61 |
MDL Number | MFCD00064552,MFCD00012632 |
InChI Key | IFQSXNOEEPCSLW-UHFFFAOYNA-N |
Synonym | dl-cysteine hydrochloride, 2-amino-3-mercaptopropanoic acid hydrochloride, cysteine hydrochloride, +--cysteine hydrochloride, dl-cysteine hcl, h-dl-cys-oh.hcl, cysteine, hydrochloride, dl, dl-cysteine hydrochloride hydrate, dl-cysteine hydrochloride anhydrous, 2-amino-3-sulfanylpropanoic acid hydrochloride |
PubChem CID | 25150 |
IUPAC Name | 2-amino-3-sulfanylpropanoic acid;hydrochloride |
SMILES | [H+].[Cl-].NC(CS)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11471130
|
Thermo Scientific Alfa Aesar
H25779.06 |
5 g |
1377.00 NOK
5g |
In Stock
Log in to see stock. |
|||||
11481130
|
Thermo Scientific Alfa Aesar
H25779.14 |
25 g | N/A | ||||||
11491130
|
Thermo Scientific Alfa Aesar
H25779.22 |
100 g |
11853.00 NOK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10318-18-0 | |
157.61 | |
IFQSXNOEEPCSLW-UHFFFAOYNA-N | |
25150 | |
[H+].[Cl-].NC(CS)C(O)=O |
C3H8ClNO2S | |
MFCD00064552,MFCD00012632 | |
dl-cysteine hydrochloride, 2-amino-3-mercaptopropanoic acid hydrochloride, cysteine hydrochloride, +--cysteine hydrochloride, dl-cysteine hcl, h-dl-cys-oh.hcl, cysteine, hydrochloride, dl, dl-cysteine hydrochloride hydrate, dl-cysteine hydrochloride anhydrous, 2-amino-3-sulfanylpropanoic acid hydrochloride | |
2-amino-3-sulfanylpropanoic acid;hydrochloride |
Specifications
140°C to 142°C | |
C3H8ClNO2S | |
4150473 | |
IFQSXNOEEPCSLW-UHFFFAOYNA-N | |
2-amino-3-sulfanylpropanoic acid;hydrochloride | |
25150 | |
≥98% | |
DL-Cysteine hydrochloride |
10318-18-0 | |
MFCD00064552,MFCD00012632 | |
dl-cysteine hydrochloride, 2-amino-3-mercaptopropanoic acid hydrochloride, cysteine hydrochloride, +--cysteine hydrochloride, dl-cysteine hcl, h-dl-cys-oh.hcl, cysteine, hydrochloride, dl, dl-cysteine hydrochloride hydrate, dl-cysteine hydrochloride anhydrous, 2-amino-3-sulfanylpropanoic acid hydrochloride | |
[H+].[Cl-].NC(CS)C(O)=O | |
157.61 | |
157.62 | |
Air sensitive; Hygroscopic |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
missing translation for 'einecsNumber' : 233-698-2
missing translation for 'rtecsNumber' : HA2200000
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only