missing translation for 'onlineSavingsMsg'
Learn More

DL-Cysteine hydrochloride, 90+%, Thermo Scientific Chemicals

Product Code. 11481130
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10318-18-0
Molecular Formula C3H8ClNO2S
Molecular Weight (g/mol) 157.61
MDL Number MFCD00064552,MFCD00012632
InChI Key IFQSXNOEEPCSLW-UHFFFAOYNA-N
Synonym dl-cysteine hydrochloride, 2-amino-3-mercaptopropanoic acid hydrochloride, cysteine hydrochloride, +--cysteine hydrochloride, dl-cysteine hcl, h-dl-cys-oh.hcl, cysteine, hydrochloride, dl, dl-cysteine hydrochloride hydrate, dl-cysteine hydrochloride anhydrous, 2-amino-3-sulfanylpropanoic acid hydrochloride
PubChem CID 25150
IUPAC Name 2-amino-3-sulfanylpropanoic acid;hydrochloride
SMILES [H+].[Cl-].NC(CS)C(O)=O

Specifications

Melting Point 140°C to 142°C
Beilstein 4150473
Formula Weight 157.62
Percent Purity ≥98%
Quantity 25 g
Sensitivity Air sensitive; Hygroscopic
Chemical Name or Material DL-Cysteine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'