Learn More
Fisherbrand™ Dispenser tips
Robust, stable, and durable dispenser tips for several brands of pipettors. For dispensing of fluids with high viscosity and high vapor pressure.
324.00 NOK - 2050.00 NOK
Specifications
Certifications/Compliance | ISO 8655 |
---|---|
For Use With (Application) | For dispensing of fluids with high viscosity, high vapor pressure |
Product Type | Dispenser Tip |
Disposable | Single-use |
Material | Polypropylene |
Product Code | Brand | Volume (Metric) | Sterility | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Sterility | Price | Quantity & Availability | |||||
15163646
|
Fisherbrand™
702609 |
0.1 mL | Non-sterile |
1090.00 NOK
Pack of 100 |
Estimated Shipment: 28-05-2025
Log in to see stock. |
|||||
15133646
|
Fisherbrand™
702610 |
0.5 mL | Non-sterile |
1040.00 NOK
Pack of 100 |
Estimated Shipment: 20-05-2025
Log in to see stock. |
|||||
15153646
|
Fisherbrand™
702612 |
1.25 mL | Non-sterile |
1250.00 NOK
Pack of 100 |
Estimated Shipment: 28-05-2025
Log in to see stock. |
|||||
15173646
|
Fisherbrand™
702614 |
2.5 mL | Non-sterile |
1040.00 NOK
Pack of 100 |
Estimated Shipment: 20-05-2025
Log in to see stock. |
|||||
15143646
|
Fisherbrand™
702616 |
5.0 mL | Non-sterile |
1040.00 NOK
Pack of 100 |
Estimated Shipment: 28-05-2025
Log in to see stock. |
|||||
15193646
|
Fisherbrand™
702618 |
12.5 mL | Non-sterile |
928.00 NOK
Pack of 100 |
Estimated Shipment: 28-05-2025
Log in to see stock. |
|||||
15183646
|
Fisherbrand™
702620 |
25 mL | Non-sterile |
639.60 NOK
Pack of 50 |
Estimated Shipment: 21-05-2025
Log in to see stock. |
|||||
15103656
|
Fisherbrand™
702622 |
50 mL | Non-sterile |
324.00 NOK
Pack of 25 |
Estimated Shipment: 20-05-2025
Log in to see stock. |
|||||
15113656
|
Fisherbrand™
702623 |
0.1 mL | Sterile |
2050.00 NOK
Pack of 100 |
Estimated Shipment: 28-05-2025
Log in to see stock. |
|||||
15123656
|
Fisherbrand™
702624 |
0.5 mL | Sterile |
1760.00 NOK
Pack of 100 |
Estimated Shipment: 28-05-2025
Log in to see stock. |
|||||
15143656
|
Fisherbrand™
702626 |
1.25 mL | Sterile |
1760.00 NOK
Pack of 100 |
Estimated Shipment: 30-05-2025
Log in to see stock. |
|||||
15133656
|
Fisherbrand™
702628 |
2.5 mL | Sterile |
1760.00 NOK
Pack of 100 |
Estimated Shipment: 30-05-2025
Log in to see stock. |
|||||
15153656
|
Fisherbrand™
702630 |
5 mL | Sterile |
1760.00 NOK
Pack of 100 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
|||||
15173656
|
Fisherbrand™
702632 |
12.5 mL | Sterile |
1760.00 NOK
Pack of 100 |
Estimated Shipment: 20-05-2025
Log in to see stock. |
|||||
15183656
|
Fisherbrand™
702636 |
50 mL | Sterile |
595.00 NOK
Pack of 25 |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
15163656
|
Fisherbrand™
702634 |
25 mL | Sterile |
605.00 NOK
Pack of 25 |
Estimated Shipment: 30-05-2025
Log in to see stock. |
|||||
Description
- These disposable tips are suitable for the following multiple dispensers:
- BRAND Handystep™ S, HandyStep™ electronic, HandyStep™ touch/ S
- Eppendorf Multipette™ 4780
- Gilson Distriman™ and Gilson Repetman™
- Ritter Ripette™ pro
- EDOS™ 5221
- Rainin AutoRep™ E, Rainin AutoRep™ S, Rainin AutoRep™ M
- STARLAB RepeatOne™
Specifications
ISO 8655 | |
Dispenser Tip | |
Polypropylene |
For dispensing of fluids with high viscosity, high vapor pressure | |
Single-use |
Product Title
Fisherbrand™ Dispenser tips