Learn More
Fisherbrand™ Dispenser tips
Robust, stable, and durable dispenser tips for several brands of pipettors. For dispensing of fluids with high viscosity and high vapor pressure.
Brand:  Fisherbrand™ 702623
1947.50 NOK valid until 2025-06-30
Use promo code "24165" to get your promotional price.
 View more versions of this product
Product Code. 15113656
  • 2050.00 NOK / Pack of 100
Fisherbrand Sweet Deals on Top Products
Use promo code 24165 at checkout
View Available Promotions
Only null left
Estimated Shipment: 09-07-2025
Add to basket
Description
Description
  • These disposable tips are suitable for the following multiple dispensers:
  • BRAND Handystep™ S, HandyStep™ electronic, HandyStep™ touch/ S
  • Eppendorf Multipette™ 4780
  • Gilson Distriman™ and Gilson Repetman™
  • Ritter Ripette™ pro
  • EDOS™ 5221
  • Rainin AutoRep™ E, Rainin AutoRep™ S, Rainin AutoRep™ M
  • STARLAB RepeatOne™
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.1 mL
ISO 8655
Dispenser Tip
Polypropylene
Sterile
For dispensing of fluids with high viscosity, high vapor pressure
Single-use
SDS
Documents Certificates
Promotions
Promotions
  • Fisherbrand
    Get up to 60% off select Fisherbrand products
    Promo Code: 24165
    Expires2025-06-30
    New Offer
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/06/2025 and 30/06/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions Save Now
Product Content Correction
Product Title
Fisherbrand™ Dispenser tips >