missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCT Mouse anti-Human, Mouse, Pig, Clone: 1B3D4, Proteintech
Mouse Monoclonal Antibody
1696.32 NOK - 4796.50 NOK
Specifications
Antigen | DCT |
---|---|
Clone | 1B3D4 |
Concentration | 1000 μg/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17721169
|
Proteintech
68114-1-IG-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17711169
|
Proteintech
68114-1-IG-150UL |
150 μL |
4796.50 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DCT Monoclonal antibody specifically detects DCT in Human, Mouse, Pig samples. It is validated for WB, ELISASpecifications
DCT | |
1000 μg/mL | |
Monoclonal | |
Liquid | |
Human, Mouse, Pig | |
1638 | |
Primary | |
Store at -20°C |
1B3D4 | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
IgG1 | |
Protein G purification | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title