missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCT Mouse anti-Human, Mouse, Pig, Clone: 1B3D4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68114-1-IG-20UL
This item is not returnable.
View return policy
Description
DCT Monoclonal antibody specifically detects DCT in Human, Mouse, Pig samples. It is validated for WB, ELISASpecifications
DCT | |
Monoclonal | |
1000 μg/mL | |
PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
Mouse | |
20 μL | |
1638 | |
Store at -20°C | |
Liquid |
Western Blot, ELISA | |
1B3D4 | |
Unconjugated | |
BC050644 | |
Protein G purification | |
Primary | |
Human, Mouse, Pig | |
Antibody | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction