Learn More
Dabigatran etexilate mesylate, MedChemExpress
MedChemExpress Dabigatran etexilate mesylate (BIBR 1048MS) is an orally active prodrug of Dabigatran (a direct inhibitor of thrombin). Dabigatran etexilate mesylate has anticoagulant effects and is used for the prophylaxis of venousthromboembolism and stroke due to atrial fibrillation.
891.99 NOK - 6425.30 NOK
Specifications
Chemical Name or Material | Dabigatran etexilate mesylate |
---|---|
CAS | 872728-81-9 |
Color | Off-White |
Purity Grade Notes | Research |
Molecular Formula | C35H45N7O8S |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16480979
|
Medchem Express
HY-10274A |
1 g |
6425.30 NOK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16480137
|
Medchem Express
HY-10274A |
50 mg |
1602.54 NOK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16460966
|
Medchem Express
HY-10274A |
200 mg |
3220.21 NOK
200mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16443758
|
Medchem Express
HY-10274A |
10 mg |
1254.83 NOK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16406487
|
Medchem Express
HY-10274A |
5 mg |
891.99 NOK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16404826
|
Medchem Express
HY-10274A |
100 mg |
2146.81 NOK
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16401556
|
Medchem Express
HY-10274A |
500 mg |
4641.34 NOK
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dabigatran etexilate mesylate (BIBR 1048MS) is an orally active prodrug of Dabigatran (a direct inhibitor of thrombin). Dabigatran etexilate mesylate has anticoagulant effects and is used for the prophylaxis of venousthromboembolism and stroke due to atrial fibrillation.Specifications
Dabigatran etexilate mesylate | |
Off-White | |
C35H45N7O8S | |
DMSO : 50 mg/mL (69.08 mM; Need ultrasonic) | |
723.84 | |
95.0% | |
Solid |
872728-81-9 | |
Research | |
BIBR 1048MS Dabigatran etexilate methanesulfonate | |
O=C(N(C1=NC=CC=C1)CCC(OCC)=O)C2=CC=C3C(N=C(N3C)CNC4=CC=C(C(NC(OCCCCCC)=O)=N)C=C4)=C2.CS(=O)(O)=O | |
723.84 | |
Research | |
COVID-19-immunoregulation |
Safety and Handling
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only