Learn More
Dabigatran etexilate mesylate, MedChemExpress
MedChemExpress Dabigatran etexilate mesylate (BIBR 1048MS) is an orally active prodrug of Dabigatran (a direct inhibitor of thrombin). Dabigatran etexilate mesylate has anticoagulant effects and is used for the prophylaxis of venousthromboembolism and stroke due to atrial fibrillation.
Brand: Medchem Express HY-10274A
This item is not returnable.
View return policy
Description
Dabigatran etexilate mesylate (BIBR 1048MS) is an orally active prodrug of Dabigatran (a direct inhibitor of thrombin). Dabigatran etexilate mesylate has anticoagulant effects and is used for the prophylaxis of venousthromboembolism and stroke due to atrial fibrillation.Specifications
Dabigatran etexilate mesylate | |
Off-White | |
C35H45N7O8S | |
BIBR 1048MS Dabigatran etexilate methanesulfonate | |
O=C(N(C1=NC=CC=C1)CCC(OCC)=O)C2=CC=C3C(N=C(N3C)CNC4=CC=C(C(NC(OCCCCCC)=O)=N)C=C4)=C2.CS(=O)(O)=O | |
723.84 | |
Research | |
COVID-19-immunoregulation |
872728-81-9 | |
Research | |
10 mg | |
DMSO : 50 mg/mL (69.08 mM; Need ultrasonic) | |
723.84 | |
95.0% | |
Solid |
Research purposes only