Learn More
Dabigatran etexilate mesylate, MedChemExpress
MedChemExpress Dabigatran etexilate mesylate (BIBR 1048MS) is an orally active prodrug of Dabigatran (a direct inhibitor of thrombin). Dabigatran etexilate mesylate has anticoagulant effects and is used for the prophylaxis of venousthromboembolism and stroke due to atrial fibrillation.
Brand:  Medchem Express HY-10274A
Encompass_Preferred
 View more versions of this product
Product Code. 16443758
  • 1254.83 NOK / 10mg
Explore more special offers
Description
Description
Dabigatran etexilate mesylate (BIBR 1048MS) is an orally active prodrug of Dabigatran (a direct inhibitor of thrombin). Dabigatran etexilate mesylate has anticoagulant effects and is used for the prophylaxis of venousthromboembolism and stroke due to atrial fibrillation.
Specifications
Specifications
Dabigatran etexilate mesylate
Off-White
C35H45N7O8S
BIBR 1048MS Dabigatran etexilate methanesulfonate
O=C(N(C1=NC=CC=C1)CCC(OCC)=O)C2=CC=C3C(N=C(N3C)CNC4=CC=C(C(NC(OCCCCCC)=O)=N)C=C4)=C2.CS(=O)(O)=O
723.84
Research
COVID-19-immunoregulation
872728-81-9
Research
10 mg
DMSO : 50 mg/mL (69.08 mM; Need ultrasonic)
723.84
95.0%
Solid