missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Serine benzyl ester hydrochloride, 95%, Thermo Scientific™
476.54 NOK - 1910.33 NOK
Chemical Identifiers
CAS | 151651-44-4 |
---|---|
Molecular Formula | C10H14ClNO3 |
Molecular Weight (g/mol) | 231.676 |
MDL Number | MFCD00191017 |
InChI Key | MGZWCDQAKCHOBX-SBSPUUFOSA-N |
Synonym | h-d-ser-obzl.hcl, h-d-ser-obzl hcl, d-serine benzyl ester hydrochloride, r-benzyl 2-amino-3-hydroxypropanoate hydrochloride, d-serinebenzylesterhydrochloride, benzyl 2r-2-amino-3-hydroxypropanoate hydrochloride, d-serine-obzl hcl, h-d-ser-obzlhcl, h-d-ser-obzl?cl, h-d-ser-obzl. hcl |
PubChem CID | 11776084 |
IUPAC Name | benzyl (2R)-2-amino-3-hydroxypropanoate;hydrochloride |
SMILES | C1=CC=C(C=C1)COC(=O)C(CO)N.Cl |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
151651-44-4 | |
231.676 | |
MGZWCDQAKCHOBX-SBSPUUFOSA-N | |
11776084 | |
C1=CC=C(C=C1)COC(=O)C(CO)N.Cl |
C10H14ClNO3 | |
MFCD00191017 | |
h-d-ser-obzl.hcl, h-d-ser-obzl hcl, d-serine benzyl ester hydrochloride, r-benzyl 2-amino-3-hydroxypropanoate hydrochloride, d-serinebenzylesterhydrochloride, benzyl 2r-2-amino-3-hydroxypropanoate hydrochloride, d-serine-obzl hcl, h-d-ser-obzlhcl, h-d-ser-obzl?cl, h-d-ser-obzl. hcl | |
benzyl (2R)-2-amino-3-hydroxypropanoate;hydrochloride |
Specifications
169°C to 174°C | |
C10H14ClNO3 | |
h-d-ser-obzl.hcl, h-d-ser-obzl hcl, d-serine benzyl ester hydrochloride, r-benzyl 2-amino-3-hydroxypropanoate hydrochloride, d-serinebenzylesterhydrochloride, benzyl 2r-2-amino-3-hydroxypropanoate hydrochloride, d-serine-obzl hcl, h-d-ser-obzlhcl, h-d-ser-obzl?cl, h-d-ser-obzl. hcl | |
C1=CC=C(C=C1)COC(=O)C(CO)N.Cl | |
231.676 | |
231.68 | |
Solid |
151651-44-4 | |
MFCD00191017 | |
MGZWCDQAKCHOBX-SBSPUUFOSA-N | |
benzyl (2R)-2-amino-3-hydroxypropanoate;hydrochloride | |
11776084 | |
95% | |
D-Serine benzyl ester hydrochloride |
Safety and Handling
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
D-Serine benzyl ester hydrochloride, 95%, Thermo Scientific™