missing translation for 'onlineSavingsMsg'
Learn More

D-Serine benzyl ester hydrochloride, 95%, Thermo Scientific™

Product Code. 15410499
5 g, 5g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151651-44-4
Molecular Formula C10H14ClNO3
Molecular Weight (g/mol) 231.676
MDL Number MFCD00191017
InChI Key MGZWCDQAKCHOBX-SBSPUUFOSA-N
Synonym h-d-ser-obzl.hcl, h-d-ser-obzl hcl, d-serine benzyl ester hydrochloride, r-benzyl 2-amino-3-hydroxypropanoate hydrochloride, d-serinebenzylesterhydrochloride, benzyl 2r-2-amino-3-hydroxypropanoate hydrochloride, d-serine-obzl hcl, h-d-ser-obzlhcl, h-d-ser-obzl?cl, h-d-ser-obzl. hcl
PubChem CID 11776084
IUPAC Name benzyl (2R)-2-amino-3-hydroxypropanoate;hydrochloride
SMILES C1=CC=C(C=C1)COC(=O)C(CO)N.Cl

Specifications

Melting Point 169°C to 174°C
Quantity 5 g
Formula Weight 231.68
Percent Purity 95%
Physical Form Solid
Chemical Name or Material D-Serine benzyl ester hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
D-Serine benzyl ester hydrochloride, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.