missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 5 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1696.32 NOK - 4796.50 NOK

Specifications

Antigen Cytokeratin 5
Concentration 767 μg/mL
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry, ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16846175
View Documents
Proteintech
28506-1-AP-20UL
20 μL
1696.32 NOK
20µL
Please to purchase this item. Need a web account? Register with us today!
16836175
View Documents
Proteintech
28506-1-AP-150UL
150 μL
4796.50 NOK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Cytokeratin 5 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Cytokeratin 5. This antibody recognizes human, mouse, rat antigen. The Cytokeratin 5 antibody has been validated for the following applications: FC, IF, IHC, WB, ELISA analysis.

Specifications

Specifications

Cytokeratin 5
Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry, ELISA
Unconjugated
Human, Rat, Mouse
IgG
767 μg/mL
Polyclonal
Rabbit
3852
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.