Learn More
Cytokeratin 5 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28506-1-AP-20UL
This item is not returnable.
View return policy
Description
The Cytokeratin 5 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Cytokeratin 5. This antibody recognizes human, mouse, rat antigen. The Cytokeratin 5 antibody has been validated for the following applications: FC, IF, IHC, WB, ELISA analysis.
Specifications
Cytokeratin 5 | |
Polyclonal | |
Unconjugated | |
20 μL | |
3852 | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry, ELISA | |
767 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |