Learn More
Cytokeratin 5 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28506-1-AP-20UL
 View more versions of this product
Product Code. 16846175
  • 1696.32 NOK / 20µL
Explore more special offers
Description
Description
The Cytokeratin 5 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Cytokeratin 5. This antibody recognizes human, mouse, rat antigen. The Cytokeratin 5 antibody has been validated for the following applications: FC, IF, IHC, WB, ELISA analysis.
Specifications
Specifications
Cytokeratin 5
Polyclonal
Unconjugated
20 μL
3852
IgG
Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry, ELISA
767 μg/mL
Rabbit
Primary
Human, Rat, Mouse