Learn More
Hettich Lab Technology™ Cyto Chamber Carrier
Hettich Lab Technology™ Cyto Chamber Carrier is suitable for holding cyto chambers for slide preparation and cell suspension examination
2195.00 NOK - 3105.00 NOK
Specifications
Type Cyto Chamber Carrier
View More Specs
Products 2
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
10553262
View Documents
Hettich Lab Technology™
1660
Rotofix 32A, Universal 320/320R, Rotina 380/380R, Rotina 420/420R Centrifuge Rotors (1726, 1748, 4758, 1624)
2195.00 NOK
Each
10557583
View Documents
Hettich Lab Technology™
5280
Rotina 380/380R, Rotina 420/420R, Rotanta 460/460R/460RC/460RF, Rotixa 500RS Centrifuge Rotors, Carriers 5051, 5268.
3105.00 NOK
Each
Description
Description
  • Suitable for cyto rotors
  • Part of the Hettich™ cyto system
Specifications
Specifications
Cyto Chamber Carrier
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Hettich Lab Technology™ Cyto Chamber Carrier