Learn More
Hettich Lab Technology™ Cyto Chamber Carrier
Hettich Lab Technology™ Cyto Chamber Carrier is suitable for holding cyto chambers for slide preparation and cell suspension examination
Brand:  Hettich Lab Technology™ 1660
 View more versions of this product
Product Code. 10553262
  • 2195.00 NOK / Each
Description
  • Suitable for cyto rotors
  • Part of the Hettich™ cyto system
Order Info
dimForUseWithEquipment
Specifications
Specifications
Cyto Chamber Carrier
Rotofix 32A, Universal 320/320R, Rotina 380/380R, Rotina 420/420R Centrifuge Rotors (1726, 1748, 4758, 1624)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Hettich Lab Technology™ Cyto Chamber Carrier > For use with: Rotofix 32A, Universal 320/320R, Rotina 380/380R, Rotina 420/420R Centrifuge Rotors (1726, 1748, 4758, 1624)