missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin T1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1696.32 NOK - 4796.50 NOK
Specifications
Antigen | Cyclin T1 |
---|---|
Concentration | 0.38 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16850585
|
Proteintech
20992-1-AP-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16840585
|
Proteintech
20992-1-AP-150UL |
150 μL |
4796.50 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The antibody is specific to CCNT1.
PAN3 is functions in cytoplasmic mRNA decay. It is as part of the Pan nuclease complex, recruits polyadenylate-binding protein which in turn stimulates PAN2 nuclease activity.Specifications
Cyclin T1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
O60563, Q9QWV9 | |
12455, 315291, 904 | |
Peptide | |
Primary | |
-20°C | |
CCNT1 |
0.38 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CCNT, CCNT1, Cyclin T, cyclin T1, CYCT1, HIVE1 | |
Ccnt1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title