missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin T1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20992-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody is specific to CCNT1.
PAN3 is functions in cytoplasmic mRNA decay. It is as part of the Pan nuclease complex, recruits polyadenylate-binding protein which in turn stimulates PAN2 nuclease activity.Specifications
Cyclin T1 | |
Polyclonal | |
Unconjugated | |
CCNT1 | |
CCNT, CCNT1, Cyclin T, cyclin T1, CYCT1, HIVE1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12455, 315291, 904 | |
-20°C | |
Liquid |
Western Blot | |
0.38 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O60563, Q9QWV9 | |
Ccnt1 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction