Learn More
Mettler Toledo™ Cyanide Half Cell Electrode
Brand:  Mettler Toledo™ 51107681
14623.47 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11977701
  • 14995.00 NOK / Each
Only null left
Estimated Shipment: 16-07-2025
Specifications
Solid State Membrane
0°C to 80°C
4 to 13
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Mettler Toledo™ Cyanide Half Cell Electrode > Membrane: Solid State