missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Culti-Loops™ Pluralibacter gergoviae ATCC™ 33028™

Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.

Brand:  Thermo Scientific™ R4601940

1859.07 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 12978938

  • 1990.00 NOK / Pack of 5

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.

The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.

Ordering Information:

Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

33028TM*
Pluralibacter gergoviae
Gram-negative Rods
5 Loops/Pack
CDC 604-77 [CIP 76.01]
Culti-Loops are CE Marked.
Single Organism
Qualitative QC
5 Loops/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number235197 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberR4601940
Lot Number164072 Certificate TypeCertificate of Analysis Date12/09/2024 Catalog NumberR4601940
Lot Number119346 Certificate TypeCertificate of Analysis Date23/05/2024 Catalog NumberR4601940
Lot Number329382 Certificate TypeCertificate of Analysis Date14/04/2024 Catalog NumberR4601940
Lot Number666594 Certificate TypeCertificate of Analysis Date04/04/2024 Catalog NumberR4601940
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Culti-Loops™ Pluralibacter gergoviae ATCC™ 33028™ > E. gergoviae ATCC 33028

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.