Learn More
Thermo Scientific™ Oxoid™ Resazurin Anaerobic Indicator
Oxoid Resazurin is an indicator strip for use with anaerobic jar systems.
Brand:  Thermo Scientific™ BR0055B
858.65 NOK valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Product Code. 10371053
  • 920.00 NOK / Pack of 100
Explore more special offers
Description
Description
Oxoid Resazurin is an indicator strip for use with anaerobic jar systems.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Anaerobic Indicator, Resazurin
Thermo Scientific Oxoid
100/Pk.
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number4417776 Certificate TypeCertificate of Analysis Date06/01/2024 Catalog NumberBR0055B
Lot Number2301138 Certificate TypeCertificate of Analysis Date06/01/2024 Catalog NumberBR0055B
Lot Number2304138 Certificate TypeCertificate of Analysis Date06/01/2024 Catalog NumberBR0055B
Lot Number2111138 Certificate TypeCertificate of Analysis Date24/08/2022 Catalog NumberBR0055B
Product Title
Thermo Scientific™ Oxoid™ Resazurin Anaerobic Indicator > 100/Pk.