Learn More
CPSF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17739-1-AP-150UL
 View more versions of this product
Product Code. 16847694
  • 4796.50 NOK / 150µL
Explore more special offers
Description
Description
CPSF2 gene ontology annotations related to this gene include mRNA cleavage; mRNA polyadenylation.
Specifications
Specifications
CPSF2
Polyclonal
Unconjugated
CPSF2
CPSF 100 kDa subunit, CPSF100, CPSF2, KIAA1367
Rabbit
Antigen Affinity Chromatography
RUO
299256, 51786, 53981
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O35218, Q9P2I0
Cpsf2
CPSF2 Fusion Protein Ag11950
150 μL
Primary
Human, Mouse, Rat