missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPSF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1696.32 NOK - 4796.50 NOK
Specifications
Antigen | CPSF2 |
---|---|
Concentration | 0.31 mg/mL |
Applications | Western Blot, Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16857694
|
Proteintech
17739-1-AP-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16847694
|
Proteintech
17739-1-AP-150UL |
150 μL |
4796.50 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CPSF2 gene ontology annotations related to this gene include mRNA cleavage; mRNA polyadenylation.Specifications
CPSF2 | |
Western Blot, Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
O35218, Q9P2I0 | |
299256, 51786, 53981 | |
CPSF2 Fusion Protein Ag11950 | |
Primary | |
-20°C | |
CPSF2 |
0.31 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CPSF 100 kDa subunit, CPSF100, CPSF2, KIAA1367 | |
Cpsf2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title