Learn More
Cole-Parmer™ CPC™ Quick Disconnect Sanitary Couplings
Integrate disposables with your existing stainless steel processing equipment.
Brand:  Cole-Parmer™ 31604-28
 View more versions of this product
Product Code. 15417383
  • 651.00 NOK / Each
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Attach these couplings directly to your sanitary hardware assemblies
  • USP Class IV materials.
  • Fully sterilizable.
  • Both couplings and inserts are available with standard sanitary terminations to attach to your system with a sanitary gasket and tri-clover clamps.
  • Choose from stainless steel or polysulfone inserts to mate to polysulfone bodies.
  • All models feature USP Class VI platinum-cured silicone gaskets.
  • Mix and match these bodies and inserts with the polysulfone quick-disconnect coupling elements for greater flexibility in your process system.
Specifications
Specifications
Sanitary x Quick-Disconnect
Polysulfone
1 in.
Quick-Disconnect, Sanitary Connector
Autoclavable
1 in.
Autoclave, e-beam, Ethylene Oxide or Gamma