missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CP110 Mouse anti-Human, Clone: 1A3C12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66448-1-IG-150UL
This item is not returnable.
View return policy
Description
GSG2 is required for normal alignment of chromosomes at metaphase.Specifications
CP110 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43303 | |
CCP110 | |
CP110 Fusion Protein Ag3489 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Western Blot | |
1A3C12 | |
Unconjugated | |
CCP110 | |
Centrosomal protein of 110 kDa, CP110, CP110 protein, KIAA0419 | |
Mouse | |
Protein A | |
RUO | |
9738 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction