missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CP110 Mouse anti-Human, Clone: 1A3C12, Proteintech
Mouse Monoclonal Antibody
1696.32 NOK - 4796.50 NOK
Specifications
Antigen | CP110 |
---|---|
Clone | 1A3C12 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16858335
|
Proteintech
66448-1-IG-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16848335
|
Proteintech
66448-1-IG-150UL |
150 μL |
4796.50 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GSG2 is required for normal alignment of chromosomes at metaphase.Specifications
CP110 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Centrosomal protein of 110 kDa, CP110, CP110 protein, KIAA0419 | |
CCP110 | |
IgG2a | |
Protein A | |
Antibody |
1A3C12 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
O43303 | |
9738 | |
CP110 Fusion Protein Ag3489 | |
Primary | |
-20°C | |
CCP110 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title