missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX4NB Mouse anti-Human, Mouse, Rat, Clone: 2B8D3, Proteintech
Mouse Monoclonal Antibody
1696.32 NOK - 4796.50 NOK
Specifications
Antigen | COX4NB |
---|---|
Clone | 2B8D3 |
Concentration | 0.5 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16848525
|
Proteintech
66547-1-IG-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16838525
|
Proteintech
66547-1-IG-150UL |
150 μL |
4796.50 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Expressed in liver, pancreas, heart, lung, kidney, brain, skeletal muscle, and placenta, Expression levels are highest in pancreas and moderate in heart, skeletal muscle, and placenta.Specifications
COX4NB | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C16orf2, C16orf4, COX4 neighbor, COX4AL, COX4NB, FAM158B, Neighbor of COX4, NOC4, Protein FAM158B | |
Emc8 | |
IgG1 | |
Protein G | |
Antibody |
2B8D3 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Mouse, Rat, Human | |
O43402, O70378, Q5FVL2 | |
10328, 18117, 361425 | |
COX4NB Fusion Protein Ag13776 | |
Primary | |
-20°C | |
EMC8 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title