missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX4NB Mouse anti-Human, Mouse, Rat, Clone: 2B8D3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66547-1-IG-20UL
This item is not returnable.
View return policy
Description
Expressed in liver, pancreas, heart, lung, kidney, brain, skeletal muscle, and placenta, Expression levels are highest in pancreas and moderate in heart, skeletal muscle, and placenta.Specifications
COX4NB | |
Monoclonal | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O43402, O70378, Q5FVL2 | |
Emc8 | |
COX4NB Fusion Protein Ag13776 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
2B8D3 | |
Unconjugated | |
EMC8 | |
C16orf2, C16orf4, COX4 neighbor, COX4AL, COX4NB, FAM158B, Neighbor of COX4, NOC4, Protein FAM158B | |
Mouse | |
Protein G | |
RUO | |
10328, 18117, 361425 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction