Learn More
Corning™ CellBIND™ 96-Well, CellBIND-Treated, Flat-Bottom Microplate
Color:Clear; Sterility:Sterile; Lid:With Lid
Brand: Corning™ 3300
This item is not returnable.
View return policy
Description
- Cell culture microplates are sterile and nonpyrogenic.
- Lids available where indicated
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Cat. No. 3841 is aseptically manufactured.
Specifications
Standard | |
0.32 cm2 | |
Corning CellBIND Surface | |
Polystyrene | |
75 to 200 μL | |
CellBIND | |
Flat Bottom (F-well) | |
50/Cs. | |
50/Cs. |
With Lid | |
8 x 12 | |
Clear | |
96 | |
Sterile | |
360 μL | |
CellBIND-treated surface helps cells adapt to reduced serum or serum-free media and improves cell attachment for increased cell growth and yield. Requires no refrigeration or special handling, unlike standard coatings used to improve cell attachment. | |
5/Pk. | |
Clear |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
COR001 Lifescience Guid
File size: 7 MB
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Special Offers
-
Corning Matrigel
,
Corning Biocoat
,
Corning CoolProduct
,
Corning Media Tech
,
Corning
Product Content Correction
Product Title
Corning™ CellBIND™ 96-Well, CellBIND-Treated, Flat-Bottom Microplate > Standard, with Lid; 5/Pk; Yes; Corning CellBIND; Flat