Learn More
Thermo Scientific™ Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
Color: Clear; Sterility: Sterile; Lid: With Lid
Brand: Thermo Scientific™ 161093
2510.00 NOK valid until 2025-12-31
Use promo code "24071" to get your promotional price.
Description
Thermo Scientific™ Nunc MicroWell 96-Well Microplates are optimized for robotics, automated readers and liquid handling systems. 96-well microplates are used for antibiotic screens, cell-based assays and screening compounds. The flat bottom shape is ideal for microscopic and optical measurements. Our plates are high quality for consistent and reliable results.
- Clear polystyrene has excellent optical properties making it ideal for optical measurements
- Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
- Untreated surfaces are ideal for adding coatings in your lab
- Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
- Plates are optimized for robotics, plate readers and liquid handlers
- Customized bar coding is available
Specifications
Custom Barcoding Available | |
With Lid | |
8 x 12 | |
Nunc | |
Microplate | |
Polystyrene | |
96 | |
Nunclon Delta | |
160 Microplates | |
160/Cs. |
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
96F well plate,Nunclon Delta, clear, with lid, Sterile (Pack of 10, Case of 160) | |
10 Pk., 160 Cases | |
Clear | |
High Flange | |
400 ÎĽL | |
Sterile | |
Flat Bottom (F-well) | |
10 Microplates |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
186098 | Certificate of Compliance | 10/06/2025 | 161093 |
186395 | Certificate of Compliance | 10/05/2025 | 161093 |
185624 | Certificate of Compliance | 09/05/2025 | 161093 |
185624 | Certificate of Irradiation | 09/05/2025 | 161093 |
185032 | Certificate of Irradiation | 21/02/2025 | 161093 |
Promotions
-
Thermo Scientific Nunc
,
Thermo Scientific Nalgene
,
Gibco
,
Invitrogen
,
Thermo Scientific
Cell Biology Promotion
Discover how you can maximise your savings and boost your research to the next level with our top-of-the-line Gibco and Thermo Scientific products.Promo Code: 24071Expires: 2025-12-31