Learn More
Ch-TOG Mouse anti-Human, Unconjugated, Clone: 1H5G1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67631-1-IG-150UL
This item is not returnable.
View return policy
Description
The Ch-TOG antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Ch-TOG. This antibody recognizes human, mouse, rat, pig antigen. The Ch-TOG antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Ch-TOG | |
Monoclonal | |
500 μg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig |
Western Blot, ELISA | |
1H5G1 | |
Unconjugated | |
150 μL | |
9793 | |
IgG1 |