missing translation for 'onlineSavingsMsg'
Learn More

Ch-TOG Mouse anti-Human, Unconjugated, Clone: 1H5G1, Proteintech

Mouse Monoclonal Antibody

1696.32 NOK - 4796.50 NOK

Specifications

Antigen Ch-TOG
Clone 1H5G1
Concentration 500 μg/mL
Applications Western Blot, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16860366
View Documents
Proteintech
67631-1-IG-20UL
20 μL
1696.32 NOK
20µL
Please to purchase this item. Need a web account? Register with us today!
16850366
View Documents
Proteintech
67631-1-IG-150UL
150 μL
4796.50 NOK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Ch-TOG antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Ch-TOG. This antibody recognizes human, mouse, rat, pig antigen. The Ch-TOG antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Specifications

Ch-TOG
500 μg/mL
Monoclonal
Mouse
9793
Primary
1H5G1
Western Blot, ELISA
Unconjugated
Human, Mouse, Rat, Pig
IgG1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.