missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ cDNA Synthesis & Go Kit

Provides a rapid and very sensitive method for first strand cDNA synthesis for use in real-time PCR studies

Brand:  MP Biomedicals™ 11EBI00005

Product Code. 15838541

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

  • cDNA Synthesis& Go Kit provides a rapid and very sensitive method for first strand cDNA synthesis for use in real-time PCR studies.
  • The 5x specific Buffer provides highly optimized components for efficient reverse transcription, and includes a unique blend of anchored oligo dT and random hexamer primers to ensure unbiased 3' and 5' coverage for enhanced data accuracy.
  • An extremely efficient reverse transcriptase delivers highly robust first strand synthesis and higher cDNA yields from a wide range of input RNA concentrations.
  • cDNA Synthesis& Go Kit offers enhanced sensitivity, efficiency and reproducibility for exceptional performance in subsequent real-time PCR experiments.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at -20°C
50 U
Gene Expression, Pathogen Detection, miRNA analysis
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MP Biomedicals™ cDNA Synthesis & Go Kit > cDNA Synthesis & go Kit, 50 rxns

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.